Learn More
R&D Systems™ MTT Cell Proliferation/Viability Assay
Safe and reliable method minimizes the number of steps necessary to assay for proliferation rate in cultured cells
Brand: R&D Systems™ 4890-025-K
Additional Details : Weight : 0.29100kg
Includes: MTT reagent; Detergent Reagent
Description
Tetrazolium compound MTT changes to from yellow to purple
MTT is added directly to the culture medium and is reduced by metabolically active cells to insoluble purple formazan dye crystals
The absorbance of the sample is read directly in the wells at an
Specifications
2500 Tests | |
MTT Cell Proliferation/Viability Assay | |
The product requires storage at 2°C to 8°C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. | |
Minimizes steps to assay for proliferation rate in cultured cells |