Learn More
R&D Systems™ MTT Cell Proliferation/Viability Assay
Safe and reliable method minimizes the number of steps necessary to assay for proliferation rate in cultured cells
£342.32 - £453.55
Specifications
Product Type | MTT Cell Proliferation/Viability Assay |
---|---|
For Use With (Application) | Minimizes steps to assay for proliferation rate in cultured cells |
Includes: MTT reagent; Detergent Reagent
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18442630
|
R&D Systems™
4890-050-K |
5000 Tests |
£453.55
5000 tests |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18441610
|
R&D Systems™
4890-025-K |
2500 Tests |
£342.32
2500 tests |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Tetrazolium compound MTT changes to from yellow to purple
MTT is added directly to the culture medium and is reduced by metabolically active cells to insoluble purple formazan dye crystals
The absorbance of the sample is read directly in the wells at an
Specifications
MTT Cell Proliferation/Viability Assay | |
Minimizes steps to assay for proliferation rate in cultured cells |