Learn More
R&D Systems™ MTT Cell Proliferation/Viability Assay
Safe and reliable method minimizes the number of steps necessary to assay for proliferation rate in cultured cells
Brand:  R&D Systems™ 4890-050-K
 View more versions of this product
Product Code. 18442630
  • £475.00 / 5000 tests
Explore more special offers
Includes: MTT reagent; Detergent Reagent
Description
Description
Tetrazolium compound MTT changes to from yellow to purple
MTT is added directly to the culture medium and is reduced by metabolically active cells to insoluble purple formazan dye crystals
The absorbance of the sample is read directly in the wells at an
Specifications
Specifications
5000 Tests
MTT Cell Proliferation/Viability Assay
The product requires storage at 2 - 8° C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Minimizes steps to assay for proliferation rate in cultured cells