Learn More
R&D Systems™ StemXVivo Serum-Free Human MSC Expansion Media
A complete media formulated and optimized for the maintenance and expansion of purified human MSCs
Brand:  R&D Systems™ CCM014
Product Code. 18431640
  • £349.00 / 500mL
Explore more special offers
Description
Description
Mesenchymal stem/stromal cells (MSCs) are a rare population of multipotent cells that can be derived from a number of tissues such as bone marrow, adipose, and placenta.
  • Lacks serum to reduce experimental variation
  • Is optimized to support reproducible human MSC growth in vitro
  • Can be supplemented with user-defined cytokines and growth factors, although these are not needed for culture
  • Is suitable for transitioning cultures to pre-clinical expansion protocols
Specifications
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Stem Cell Expansion Media
500 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ StemXVivo Serum-Free Human MSC Expansion Media > 500mL