Learn More
(R)-(-)-S-Methyl-S-phenylsulfoximine, 97%, Thermo Scientific Chemicals
£50.15 - £144.72
Chemical Identifiers
CAS | 60933-65-5 |
---|---|
Molecular Formula | C7H9NOS |
Molecular Weight (g/mol) | 155.215 |
MDL Number | MFCD00151460 |
InChI Key | YFYIDTVGWCYSEO-JTQLQIEISA-N |
Synonym | s-+-s-methyl-s-phenylsulfoximine, s-+-s-methyl-s-phenylsulfoximine ee, s-imino methyl phenyl-??-sulfanone, r---s-methyl-s-phenylsulfoximine ee, imino-methyl-oxo-phenyl-$l^ 6, s-+-s-methyl-s-phenylsulphoximine, sulfoximine,s-methyl-s-phenyl-, s s |
PubChem CID | 25036288 |
IUPAC Name | imino-methyl-oxo-phenyl-$l^{6}-sulfane |
SMILES | CS(=N)(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11342586
|
Thermo Scientific Alfa Aesar
L08562.MD |
250 mg |
£50.15
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11332586
|
Thermo Scientific Alfa Aesar
L08562.03 |
1 g |
£144.72
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(R)-(-)-S-Methyl-S-phenylsulfoximine is used as chiral ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(R)-(-)-S-Methyl-S-phenylsulfoximine is used as chiral ligands.
Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with strong oxidizing agents.
Chemical Identifiers
60933-65-5 | |
155.215 | |
YFYIDTVGWCYSEO-JTQLQIEISA-N | |
25036288 | |
CS(=N)(=O)C1=CC=CC=C1 |
C7H9NOS | |
MFCD00151460 | |
s-+-s-methyl-s-phenylsulfoximine, s-+-s-methyl-s-phenylsulfoximine ee, s-imino methyl phenyl-??-sulfanone, r---s-methyl-s-phenylsulfoximine ee, imino-methyl-oxo-phenyl-$l^ 6, s-+-s-methyl-s-phenylsulphoximine, sulfoximine,s-methyl-s-phenyl-, s s | |
imino-methyl-oxo-phenyl-$l^{6}-sulfane |
Specifications
60933-65-5 | |
C7H9NOS | |
5245635 | |
s-+-s-methyl-s-phenylsulfoximine, s-+-s-methyl-s-phenylsulfoximine ee, s-imino methyl phenyl-??-sulfanone, r---s-methyl-s-phenylsulfoximine ee, imino-methyl-oxo-phenyl-$l^ 6, s-+-s-methyl-s-phenylsulphoximine, sulfoximine,s-methyl-s-phenyl-, s s | |
−36° (c=1 in Acetone) | |
imino-methyl-oxo-phenyl-$l^{6}-sulfane | |
25036288 | |
97% |
29°C to 31°C | |
MFCD00151460 | |
Hygroscopic | |
YFYIDTVGWCYSEO-JTQLQIEISA-N | |
CS(=N)(=O)C1=CC=CC=C1 | |
155.215 | |
155.22 | |
(R)-(-)-S-Methyl-S-phenylsulfoximine |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.