Learn More
Tocris™ (R)-ZINC 3573
MRGPRX2 agonist
Brand:  Tocris™ 6351/5
 View more versions of this product
Product Code. 18778036
  • £82.07 / 5mg
Explore more special offers
Description
Description
(R)-ZINC 3573 is a Mas-related G protein-coupled receptor X2 (MRGPRX2) agonist (EC50 = 0.74 μM), which displays selectivity for MRGPRX2 over 350 other GPCRs, including MRGPRX1. (R)-ZINC 3573 induces intracellular calcium release and degranulation in LAD2 mast cells in vitro.
Negative control (S)-ZINC 3573 (Cat. No. 6352) also available.
Specifications
Specifications
5 mg
307.4
98%
C18H21N5
Agonist