Learn More
Tocris™ (R)-ZINC 3573
MRGPRX2 agonist
£82.07 - £315.33
Specifications
Molecular Formula | C18H21N5 |
---|---|
Formula Weight | 307.4 |
Product Type | Agonist |
Purity | 98% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18778036
|
Tocris™
6351/5 |
5 mg |
£82.07
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18756076
|
Tocris™
6351/25 |
25 mg |
£315.33
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(R)-ZINC 3573 is a Mas-related G protein-coupled receptor X2 (MRGPRX2) agonist (EC50 = 0.74 μM), which displays selectivity for MRGPRX2 over 350 other GPCRs, including MRGPRX1. (R)-ZINC 3573 induces intracellular calcium release and degranulation in LAD2 mast cells in vitro.Negative control (S)-ZINC 3573 (Cat. No. 6352) also available.
Specifications
C18H21N5 | |
307.4 | |
Agonist | |
98% |
Safety and Handling
Recommended Storage : Store at 4°C