missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat ALK-7 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF577
This item is not returnable.
View return policy
Description
ALK-7/Activin Receptor Type 1C Polyclonal specifically detects ALK-7/Activin Receptor Type 1C in Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ALK-7/Activin Receptor Type 1C | |
Polyclonal | |
Unconjugated | |
NP_620790 | |
ACVR1C | |
Mouse myeloma cell line NS0-derived recombinant rat ALK-7 Leu26-Arg108 (predicted) Accession # NP_620790 | |
100 μg | |
Primary | |
Detects rat ALK-7 in direct ELISAs and Western blots. | |
Rat | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
activin A receptor, type IC, Activin receptor type IC, activin receptor type-1C, Activin receptor-like kinase 7, Activin RIC, ACTR-IC, ACVR1C, ACVRLK7, ALK7, ALK7ALK-7, EC 2.7.11, EC 2.7.11.30 | |
Goat | |
Affinity Purified | |
RUO | |
130399 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only