missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat ALK-7 Antibody, R&D Systems™
Goat Polyclonal Antibody
£146.86 - £393.97
Specifications
Antigen | ALK-7/Activin Receptor Type 1C |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18725517
|
R&D Systems
AF577-SP |
25 μg |
£146.86
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18001422
|
R&D Systems
AF577 |
100 μg |
£393.97
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALK-7/Activin Receptor Type 1C Polyclonal specifically detects ALK-7/Activin Receptor Type 1C in Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ALK-7/Activin Receptor Type 1C | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Rat | |
activin A receptor, type IC, Activin receptor type IC, activin receptor type-1C, Activin receptor-like kinase 7, Activin RIC, ACTR-IC, ACVR1C, ACVRLK7, ALK7, ALK7ALK-7, EC 2.7.11, EC 2.7.11.30 | |
ACVR1C | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_620790 | |
130399 | |
Mouse myeloma cell line NS0-derived recombinant rat ALK-7 Leu26-Arg108 (predicted) Accession # NP_620790 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects rat ALK-7 in direct ELISAs and Western blots. |
For Research Use Only