missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Rat IL-15 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 400-24-250UG
This item is not returnable.
View return policy
Description
IL-15 (Interleukin 15 ) is a cytokine that regulates T and natural killer cell activation and proliferation. IL-15 and IL-2 share many biological activities as both have been found to bind common hematopoietin receptor subunits, and may compete for the same receptor, and thus negatively regulate each other's activity. The number of CD8+ memory cells is shown to be controlled by a balance between IL-15 and IL-2. IL-15 induces the activation of JAK kinases, as well as the phosphorylation and activation of transcription activators STAT3, STAT5, and STAT6. In mouse, studies suggest that IL-15 may increase the expression of apoptosis inhibitor Bcl-xL, possibly through the transcription activation activity of STAT6, and thus prevent apoptosis. IL-15 plays an important role in the growth and differentiation of T and B lymphocytes, natural killer cells, macrophages, and monocytes as well as activation of a number of important intracellular signaling molecules. As such, IL-15 could be essential for the immune responses, allograft rejection, and the pathogenesis of autoimmune diseases. Further, IL-15 is a widely expressed pro-inflammatory cytokine and has been shown to play a role in several inflammatory disorders, including rheumatoid arthritis, psoriasis and pulmonary inflammatory diseases. Emerging data suggest that IL-15 may serve as a good therapeutic target, as there appears to be a beneficial effect of IL-15 neutralization in models of psoriasis and diabetes.Specifications
P97604 | |
protein with no preservative | |
13.5 | |
250 μg | |
-20°C | |
<1 EU/μg | |
IL15 | |
Unconjugated | |
MNWIDVRYDL EKIESLIQFI HIDTTLYTDS DFHPSCKVTA MNCFLLELQV ILHEYSNMTL NETVRNVLYL ANSTLSSNKN VIESGCKECE ELEERNFTEF LQSFIHIVQM FINTS | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
25670 | |
Rat IL-15 | |
E. coli | |
RUO | |
AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15 | |
Rat | |
Recombinant | |
Lyophilized |