missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Rat (Sprague-Dawley) Cryopreserved Hepatocytes, Plateable Male

Rat (Sprague-Dawley) Cryopreserved Hepatocytes, Plateable Male

Brand:  Gibco™ RTCP10

Product Code. 10796411

  • £477.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

These plateable cryopreserved male rat (Sprague-Dawley) hepatocytes have been characterized for general Phase I and Phase II drug metabolizing enzyme activities.

Cells per vial: 4-8 million

Get the Results You Need with GIBCO Hepatocytes
High quality hepatocytes, with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure you receive the highest quality cryopreserved hepatocytes.

Each lot is tested for:
  • Phase I and II enzyme activities
  • Proper morphology
  • Viabilities generally >75%
  • Attachment efficiency
  • Monolayer formation and integrity

    For research use only. Not intended for human or animal therapeutic or diagnostic use.

    Related Links:

    Learn more about our cryopreserved hepatocytes

    Learn about our full line of hepatocytes ,

    Note:
    These products require shipment in LN2 vapor dewars therefore additional fees may apply.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Store in a cryostorage dewar or -135°C freezer.
    Cryopreserved
    Rat
    4-8 x 106
    Hepatocytes
    Hepatocytes
    Male
    Pooled
    Gibco™
    1.5 mL
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents Certificates

    Certificates

    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot NumberRS1003 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberRTCP10
    Lot NumberRS1006 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberRTCP10
    Lot NumberRS1007 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberRTCP10
    Lot NumberRS1008 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberRTCP10
    Lot NumberRS1009 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberRTCP10
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Gibco™ Rat (Sprague-Dawley) Cryopreserved Hepatocytes, Plateable Male > 1.5mL

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.