missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Rat (Sprague Dawley) Microsomes
Rat (Sprague Dawley) Microsomes
Brand: Gibco™ RTMCPL
Code : 50
Additional Details : Weight : 0.50000kg
Description
Liver microsomes prepared from a pool of male Sprague Dawley rats.Total protein concentration: 20 mg/ml
We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.
For research use only. Not intended for human or animal therapeutic or diagnostic use.
Related Link
Learn more about our microsomes and subcellular fractions
Specifications
20 mg/mL | |
Store between -60°C and -85°C | |
Male | |
Microsomes | |
Liver | |
Gibco™ |
Subcellular Fractions | |
Pooled | |
Dry Ice | |
Rat | |
Frozen | |
0.5 mL |
For Research Use Only. Not for use in diagnostic procedures.