Learn More
Lenz Laborglasintrumente Reaction Vessels, Spherical, with Withdrawal Valve
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 06008488
 View more versions of this product
Product Code. 11542234
  • £1815.00 / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Without graduation
  • With laboratory flange LF
  • With groove
  • Made of DURAN™ tubing
  • Other sizes on request
Specifications
Specifications
10 L
430 mm
280 mm
Duran Glass
Lab Flange 150 LF