Learn More
Lenz Laborglasintrumente Reaction Vessels, Spherical, with Withdrawal Valve
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
£673.00 - £2480.00
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Includes | Price | Quantity & Availability | |||
11502234
|
Lenz Laborglasintrumente
06008285 |
6 L | 236 mm | Lab Flange 100 LF |
£959.00
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
11512234
|
Lenz Laborglasintrumente
06008288 |
10 L | 280 mm | Lab Flange 100 LF |
£1635.00
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
11522234
|
Lenz Laborglasintrumente
06008294 |
20 L | 350 mm | Lab Flange 100 LF |
£2185.00
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
11532234
|
Lenz Laborglasintrumente
06008485 |
6 L | 236 mm | Lab Flange 150 LF |
£1095.00
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
11542234
|
Lenz Laborglasintrumente
06008488 |
10 L | 280 mm | Lab Flange 150 LF |
£1815.00
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
11552224
|
Lenz Laborglasintrumente
06008149 |
0.25 L | 85 mm | Lab Flange 60 LF |
£673.00
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
11552234
|
Lenz Laborglasintrumente
06008494 |
20 L | 350 mm | Lab Flange 150 LF |
£2335.00
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
11562224
|
Lenz Laborglasintrumente
06008158 |
0.5 L | 105 mm | Lab Flange 60 LF |
£681.00
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
11562234
|
Lenz Laborglasintrumente
06008585 |
6 L | 236 mm | Lab Flange 200 LF |
£1250.00
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
11572224
|
Lenz Laborglasintrumente
06008170 |
1 L | 131 mm | Lab Flange 60 LF |
£695.00
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
11572234
|
Lenz Laborglasintrumente
06008588 |
10 L | 280 mm | Lab Flange 200 LF |
£1965.00
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
11582224
|
Lenz Laborglasintrumente
06008273 |
2 L | 165 mm | Lab Flange 100 LF |
£732.00
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
11582234
|
Lenz Laborglasintrumente
06008594 |
20 L | 350 mm | Lab Flange 200 LF |
£2480.00
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
11592224
|
Lenz Laborglasintrumente
06008279 |
4 L | 206 mm | Lab Flange 100 LF |
£866.00
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
Description
- Without graduation
- With laboratory flange LF
- With groove
- Made of DURAN™ tubing
- Other sizes on request
Specifications
Duran Glass |