Learn More
Thermo Scientific™ Reagent Reservoirs
GREENER_CHOICE
Choose these reagent reservoirs, designed to provide a variety of solutions to suit all of your pipetting procedures.
£27.00 - £337.00
Products 4
Product Code Brand Capacity (Metric) Material Sterility Packaging Price Quantity & Availability  
Product Code Brand Capacity (Metric) Material Sterility Packaging Price Quantity & Availability  
11475748
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
95128095
25 mL Polystyrene Non-sterile 10 Bags of 10
£331.00
Pack of 100
In Stock
to see stock.
Edge
Add to basket
10125707
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
95128085
100 mL Polystyrene Non-sterile 10 Bags of 10
£337.00
Pack of 100
In Stock
to see stock.
Edge
Add to basket
10481121
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
9510027
60 mL Polypropylene Non-sterile Bulk
£27.00
Pack of 5
In Stock
to see stock.
Edge
Add to basket
11405758
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
95128093
25 mL Polystyrene Non-sterile Bulk
£309.00
Pack of 100
In Stock
to see stock.
Edge
Add to basket
Description
Description
Thermo Scientific™ Reagent Reservoirs provide a variety of solutions to suit all of your pipetting procedures.
Innovation goes beyond our pipetters and tips. Even our reagent reservoirs have been designed to provide solutions tailored to your procedures, rather than the other way around. Four variations of reservoirs are offered, 25mL, 25mL with divider, 100mL and a disposable polystyrene 75mL reservoir.
Benefits
  • Trough within a trough maximizes the amount of liquid accessible to pipet tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Extra-wide base adds rigidity and stability to reservoir, helping to avoid spills
  • 25mL Divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizes recovery if using only single channel and small amount of reagent
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Reagent Reservoirs