missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Rec Protein A

Rec Protein A

Brand:  Novex™ 101100

Code : 50

Additional Details : Weight : 0.02100kg

Product Code. 10029462

  • £98.75 / 5mg
Estimated Shipment: 05-08-2024
to see stock.
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

This recombinant protein A from Staphylococcus aureus Cowan I was expressed in Bacillus. The gene has been altered to truncate the protein A at the carboxyl terminal. This does not affect the binding characteristics.
Specifications

Specifications

Store at 2°C to 8°C
Protein A
5 mg
Wet Ice
Unconjugated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.