Learn More
Velp Scientifica™ JP Recirculating Water Pump
Ensures optimum aspiration depending on the digestion phases and the number of samples placed in the digester connected to the pump. Velp Scientifica™ JP Recirculating Water Pumps eliminate the problem of unreliable water supply.
Brand:  Velp Scientifica™
Code : NEW
Additional Details : Weight : 8.40000kg
 View more versions of this product
Product Code. 11311620
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Aspiration matches digestion phases and sample number
  • Up to 35 liters/minute airflow pumping speed
  • Selectable performance mode related to samples
  • Chemically resistant ABS structure
  • Premium quality materials
  • Water recirculated for economy
  • Quiet operation
  • Water level detection
  • Convenient drain valve
  • Easy to move using two inset handles
Specifications
Specifications
Recirculating Water Pump
60 Hz
230 V
ABS