Learn More
Velp Scientifica™ JP Recirculating Water Pump
Ensures optimum aspiration depending on the digestion phases and the number of samples placed in the digester connected to the pump. Velp Scientifica™ JP Recirculating Water Pumps eliminate the problem of unreliable water supply.
£1130.00
Product Code | Brand | Hertz | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Hertz | Price | Quantity & Availability | |||||
11751164
|
Velp Scientifica™
F30620198 |
50 Hz |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11311620
|
Velp Scientifica™
F30630198 |
60 Hz |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Aspiration matches digestion phases and sample number
- Up to 35 liters/minute airflow pumping speed
- Selectable performance mode related to samples
- Chemically resistant ABS structure
- Premium quality materials
- Water recirculated for economy
- Quiet operation
- Water level detection
- Convenient drain valve
- Easy to move using two inset handles
Specifications
Recirculating Water Pump | |
230 V | |
ABS |