Learn More
Velp Scientifica™ JP Recirculating Water Pump
Ensures optimum aspiration depending on the digestion phases and the number of samples placed in the digester connected to the pump. Velp Scientifica™ JP Recirculating Water Pumps eliminate the problem of unreliable water supply.
Brand: Velp Scientifica™ F30620198
Code : A0
Additional Details : Weight : 10.90000kg
Description
- Aspiration matches digestion phases and sample number
- Up to 35 liters/minute airflow pumping speed
- Selectable performance mode related to samples
- Chemically resistant ABS structure
- Premium quality materials
- Water recirculated for economy
- Quiet operation
- Water level detection
- Convenient drain valve
- Easy to move using two inset handles
Specifications
Recirculating Water Pump | |
50 Hz | |
230 V | |
ABS |