missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine L2HGDH, mitochondrial Protein
A DNA sequence encoding the Bos taurus (Bovine) L2HGDH, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6285-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_001094560.1 | |
L2HGDH, mitochondrial Protein | |
Research Use Only | |
L2HGDH | |
Recombinant Protein | |
E. coli | |
IIGGGIVGLA SARALILRHP ALSIGVLEKE KNLAVHQTGH NSGVIHSGIY YKPESLKAKL CVQGAALIYE YCNQKGISYK QCGKLIVAVE QEEIPRLQAL YERGLQNGVQ GLRLIQQEDI KKKEPYCRGL MAIDCPYTGI VDYRQVAFSF AKDFQEAGGS VLTNFEVEDI EMARESPSRS KDGMKYPIVI RNTKGEEVRC QYVVTCAGLY SDRISELSGC NPNPRIVPFR GDYLVLKPEK RYLVKGNIYP VPDSRFPFLG VHFTPRMDGN IWLGPNAILA FKREGYRPFD FSARDIMDII IKSGLIKLVF QNFSYGVNEM YKACFLSATV KHLQKFIPEI TISDVLRGPA GVRAQALDRD GNLIEDFVFD GGVGDIGNRI LHVRNAPSPA ATSSLAISGM IADEVQQRFK L | |
Greater than 90% as determined by SDS-PAGE. |
514230 | |
100 μg | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Bovine L2HGDH, mitochondrial Protein > 100μg
Spot an opportunity for improvement?Share a Content Correction