missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Carbon storage regulator Protein
A DNA sequence encoding the Escherichia coli (strain K12 / DH10B) Carbon storage regulator, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | WP_000906486.1 |
---|---|
Gene ID (Entrez) | 32441215 |
Name | Carbon storage regulator Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16009100
|
enQuireBio™
QP7510-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 02-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16029100
|
enQuireBio™
QP7510-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 02-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16019100
|
enQuireBio™
QP7510-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16039100
|
enQuireBio™
QP7510-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049100
|
enQuireBio™
QP7510-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16059100
|
enQuireBio™
QP7510-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_000906486.1 | |
Carbon storage regulator Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
32441215 | |
Research Use Only | |
csrA | |
Recombinant Protein | |
E. coli | |
MLILTRRVGE TLMIGDEVTV TVLGVKGNQV RIGVNAPKEV SVHREEIYQR IQAEKSQQSS Y | |
Greater than 90% as determined by SDS-PAGE. |