missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli DNA ligase Protein
A DNA sequence encoding the Escherichia coli (strain K12) DNA ligase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7075-200ug
This item is not returnable.
View return policy
Specifications
NP_416906.1 | |
DNA ligase Protein | |
Research Use Only | |
ligA | |
Recombinant Protein | |
E. coli | |
MESIEQQLTE LRTTLRHHEY LYHVMDAPEI PDAEYDRLMR ELRELETKHP ELITPDSPTQ RVGAAPLAAF SQIRHEVPML SLDNVFDEES FLAFNKRVQD RLKNNEKVTW CCELKLDGLA VSILYENGVL VSAATRGDGT TGEDITSNVR TIRAIPLKLH GENIPARLEV RGEVFLPQAG FEKINEDARR TGGKVFANPR NAAAGSLRQL DPRITAKRPL TFFCYGVGVL EGGELPDTHL GRLLQFKKWG LPVSDRVTLC ESAEEVLAFY HKVEEDRPTL GFDIDGVVIK VNSLAQQEQL GFVARAPRWA VAFKFPAQEQ MTFVRDVEFQ VGRTGAITPV ARLEPVHVAG VLVSNATLHN ADEIERLGLR IGDKVVIRRA GDVIPQVVNV VLSERPEDTR EVVFPTHCPV CGSDVERVEG EAVARCTGGL ICGAQRKESL KHFVSRRAMD VDGMGDKIID QLVEKEYVHT PADLFKLTAG KLTGLERMGP KSAQNVVNAL EKAKETTFAR FLYALGIREV GEATAAGLAA YFGTLEALEA ASIEELQKVP DVGIVVASHV HNFFAEESNR NVISELLAEG VHWPAPIVIN AEEIDSPFAG KTVVLTGSLS QMSRDDAKAR LVELGAKVAG SVSKKTDLVI AGEAAGSKLA KAQELGIEVI DEAEMLRLLG S | |
Greater than 90% as determined by SDS-PAGE. |
946885 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |