missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli DNA ligase Protein
A DNA sequence encoding the Escherichia coli (strain K12) DNA ligase, was expressed in the hosts and tags indicated.
£320.00 - £2725.00
Specifications
Accession Number | NP_416906.1 |
---|---|
Gene ID (Entrez) | 946885 |
Name | DNA ligase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047290
|
enQuireBio™
QP7075-10UG |
10 μg |
£320.00
10µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087290
|
enQuireBio™
QP7075-50UG |
50 μg |
£458.00
50µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037290
|
enQuireBio™
QP7075-100UG |
100 μg |
£780.00
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067290
|
enQuireBio™
QP7075-200UG |
200 μg |
£1285.00
200µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077290
|
enQuireBio™
QP7075-500UG |
500 μg |
£1715.00
500µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057290
|
enQuireBio™
QP7075-1MG |
1 mg |
£2725.00
1mg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_416906.1 | |
DNA ligase Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
946885 | |
Research Use Only | |
ligA | |
Recombinant Protein | |
E. coli | |
MESIEQQLTE LRTTLRHHEY LYHVMDAPEI PDAEYDRLMR ELRELETKHP ELITPDSPTQ RVGAAPLAAF SQIRHEVPML SLDNVFDEES FLAFNKRVQD RLKNNEKVTW CCELKLDGLA VSILYENGVL VSAATRGDGT TGEDITSNVR TIRAIPLKLH GENIPARLEV RGEVFLPQAG FEKINEDARR TGGKVFANPR NAAAGSLRQL DPRITAKRPL TFFCYGVGVL EGGELPDTHL GRLLQFKKWG LPVSDRVTLC ESAEEVLAFY HKVEEDRPTL GFDIDGVVIK VNSLAQQEQL GFVARAPRWA VAFKFPAQEQ MTFVRDVEFQ VGRTGAITPV ARLEPVHVAG VLVSNATLHN ADEIERLGLR IGDKVVIRRA GDVIPQVVNV VLSERPEDTR EVVFPTHCPV CGSDVERVEG EAVARCTGGL ICGAQRKESL KHFVSRRAMD VDGMGDKIID QLVEKEYVHT PADLFKLTAG KLTGLERMGP KSAQNVVNAL EKAKETTFAR FLYALGIREV GEATAAGLAA YFGTLEALEA ASIEELQKVP DVGIVVASHV HNFFAEESNR NVISELLAEG VHWPAPIVIN AEEIDSPFAG KTVVLTGSLS QMSRDDAKAR LVELGAKVAG SVSKKTDLVI AGEAAGSKLA KAQELGIEVI DEAEMLRLLG S | |
Greater than 90% as determined by SDS-PAGE. |