missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli F17 fimbrial Protein

A DNA sequence encoding the Escherichia coli (strain K12) F17 fimbrial protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7022-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16077080

  • £320.00 / 10µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

WP_061363423.1
10 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
F17 fimbrial Protein
Research Use Only
F17a-A
Recombinant Protein
E. coli
YDGKITFNGK VVDQTCSVTT ESKNLTVKLP TVSANSLASS GKVVGLTPFT ILLEGCNTPA VTGAQNVNAY FEPNANTDYT TGNLTNTASS GASNVQIQLL NADGVKAIKL GQAAAAQSVD TVAINDANVT LRYNAQYYAT GVATAGDVTS TVNYTIAYQ
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers