missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli F17 fimbrial Protein
A DNA sequence encoding the Escherichia coli (strain K12) F17 fimbrial protein, was expressed in the hosts and tags indicated.
£320.00 - £2725.00
Specifications
Accession Number | WP_061363423.1 |
---|---|
Name | F17 fimbrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | F17a-A |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16077080
|
enQuireBio™
QP7022-10UG |
10 μg |
£320.00
10µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017090
|
enQuireBio™
QP7022-50UG |
50 μg |
£458.00
50µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067080
|
enQuireBio™
QP7022-100UG |
100 μg |
£780.00
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16097080
|
enQuireBio™
QP7022-200UG |
200 μg |
£1285.00
200µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007090
|
enQuireBio™
QP7022-500UG |
500 μg |
£1715.00
500µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087080
|
enQuireBio™
QP7022-1MG |
1 mg |
£2725.00
1mg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
WP_061363423.1 | |
Research Use Only | |
F17a-A | |
Recombinant Protein | |
E. coli | |
YDGKITFNGK VVDQTCSVTT ESKNLTVKLP TVSANSLASS GKVVGLTPFT ILLEGCNTPA VTGAQNVNAY FEPNANTDYT TGNLTNTASS GASNVQIQLL NADGVKAIKL GQAAAAQSVD TVAINDANVT LRYNAQYYAT GVATAGDVTS TVNYTIAYQ | |
Greater than 90% as determined by SDS-PAGE. |
F17 fimbrial Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |