missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli F17 fimbrial Protein
A DNA sequence encoding the Escherichia coli (strain K12) F17 fimbrial protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7022-200ug
Additional Details : Weight : 0.01000kg
Specifications
WP_061363423.1 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
F17 fimbrial Protein | |
Research Use Only | |
F17a-A | |
Recombinant Protein | |
E. coli | |
YDGKITFNGK VVDQTCSVTT ESKNLTVKLP TVSANSLASS GKVVGLTPFT ILLEGCNTPA VTGAQNVNAY FEPNANTDYT TGNLTNTASS GASNVQIQLL NADGVKAIKL GQAAAAQSVD TVAINDANVT LRYNAQYYAT GVATAGDVTS TVNYTIAYQ | |
Greater than 90% as determined by SDS-PAGE. |