missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Lipopolysaccharide assembly protein 1
A DNA sequence encoding the Escherichia coli (strain K12) Lipopolysaccharide assembly protein B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7327-ec-50ug
This item is not returnable.
View return policy
Specifications
WP_000891353.1 | |
50 μg | |
Not determined. | |
Cytoplasmic Domain | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Lipopolysaccharide assembly protein 1 | |
Research Use Only | |
yciM | |
Recombinant Protein | |
E. coli | |
WYMGRRSAQQ NKQDEANRLS RDYVAGVNFL LSNQQDKAVD LFLDMLKEDT GTVEAHLTLG NLFRSRGEVD RAIRIHQTLM ESASLTYEQR LLAIQQLGRD YMAAGLYDRA EDMFNQLTDE TDFRIGALQQ LLQIYQATSE WQKAIDVAER LVKLGKDKQR VEIAHFYCEL ALQHMASDDL DRAMTLLKKG AAADKNSARV SIMMGRVFMA KGEYAKAVES LQRVISQDRE LVSETLEMLQ TCYQQLGKTA EWAEFLQRAV EENTGADAEL MLADIIEARD GSEAAQVYIT RQLQRHPTMR VFHKLMDYHL NEAEEGRAKE SLMVLRDMVG EKVRSKPRYR CQKCGFTAYT LYWHCPSCRA WSTIKPIRGL DGL | |
Greater than 90% as determined by SDS-PAGE. |