missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ASCC3 Protein
A DNA sequence encoding the Homo sapiens (Human) ASCC3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5683-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_001271200.1 | |
ASCC3 Protein | |
Research Use Only | |
ASCC3 | |
Recombinant Protein | |
E. coli | |
MALPRLTGAL RSFSNVTKQD NYNEEVADLK IKRSKLHEQV LDLGLTWKKI IKFLNEKLEK SKMQSINEDL KDILHAAKQI EVNCPFQKRR LDGKEEDEKM SRASDRFRGL R | |
Greater than 80% as determined by SDS-PAGE. |
10973 | |
50 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |