missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP synthase subunit O, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) ATP synthase subunit O, mitochondrial, was expressed in the hosts and tags indicated.
Specifications
Accession Number | NP_001688.1 |
---|---|
Gene ID (Entrez) | 539 |
Name | ATP synthase subunit O, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16074611
|
enQuireBio™
QP8437-ec-10ug |
10 μg |
N/A
|
N/A | |||||
16014621
|
enQuireBio™
QP8437-ec-50ug |
50 μg |
N/A
|
N/A | |||||
16064611
|
enQuireBio™
QP8437-ec-100ug |
100 μg |
N/A
|
N/A | |||||
16094611
|
enQuireBio™
QP8437-ec-200ug |
200 μg |
N/A
|
N/A | |||||
16004621
|
enQuireBio™
QP8437-ec-500ug |
500 μg |
N/A
|
N/A | |||||
16084611
|
enQuireBio™
QP8437-ec-1mg |
1 mg |
N/A
|
N/A | |||||
Specifications
NP_001688.1 | |
ATP synthase subunit O, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
539 | |
Research Use Only | |
ATP5O | |
Recombinant Protein | |
E. coli | |
FAKLVRPPVQ VYGIEGRYAT ALYSAASKQN KLEQVEKELL RVAQILKEPK VAASVLNPYV KRSIKVKSLN DITAKERFSP LTTNLINLLA ENGRLSNTQG VVSAFSTMMS VHRGEVPCTV TSASPLEEAT LSELKTVLKS FLSQGQVLKL EAKTDPSILG GMIVRIGEKY VDMSVKTKIQ KLGRAMREIV | |
Greater than 90% as determined by SDS-PAGE. |