Learn More
R&D Systems™ Recombinant Human BMP-5 (CHO-expressed) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 615-BMC-020/CF
 View more versions of this product
Product Code. 18127960
  • £350.96 / 20µg
Explore more special offers
Description
Description
The Recombinant Human BMP-5 (CHO-expressed) Protein is derived from CHO. The Recombinant Human BMP-5 (CHO-expressed) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in HCl.
15.6 kDa
20μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -70° C under sterile conditions after reconstitution.
Recombinant
653
BMP-5
Chinese Hamster Ovary cell line,CHO-derived human BMP-5 protein Ala317-His454
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.