missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human BMP-5 (CHO-expressed) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£350.96
Specifications
Gene ID (Entrez) | 653 |
---|---|
Molecular Weight (g/mol) | 15.6 kDa |
Name | BMP-5 |
Quantity | 20μg |
Source | Chinese Hamster Ovary cell line,CHO-derived human BMP-5 protein Ala317-His454 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18127960
|
R&D Systems™
615-BMC-020/CF |
Lyophilized from a 0.2μm filtered solution in HCl. |
£350.96
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18198130
|
R&D Systems™
615-BMC-020 |
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. |
£350.96
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human BMP-5 (CHO-expressed) Protein is derived from CHO. The Recombinant Human BMP-5 (CHO-expressed) Protein has been validated for the following applications: Bioactivity.Specifications
653 | |
BMP-5 | |
Chinese Hamster Ovary cell line,CHO-derived human BMP-5 protein Ala317-His454 | |
Recombinant |
15.6 kDa | |
20μg | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |