Learn More
R&D Systems™ Recombinant Human BMP-5 (CHO-expressed) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 615-BMC-020
This item is not returnable.
View return policy
Description
The Recombinant Human BMP-5 (CHO-expressed) Protein is derived from CHO. The Recombinant Human BMP-5 (CHO-expressed) Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. | |
15.6 kDa | |
20μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
653 | |
BMP-5 | |
Chinese Hamster Ovary cell line,CHO-derived human BMP-5 protein Ala317-His454 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |