missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Carbonic Anhydrase XII / CA12 Protein

A DNA sequence encoding the Homo sapiens (Human) Carbonic Anhydrase XII / CA12, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5742-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15957439

  • £467.00 / 100µg
Estimated Shipment: 02-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_001209.1
Carbonic Anhydrase XII / CA12 Protein
Research Use Only
CA12
Recombinant Protein
Yeast
APVNGSKWTY FGPDGENSWS KKYPSCGGLL QSPIDLHSDI LQYDASLTPL EFQGYNLSAN KQFLLTNNGH SVKLNLPSDM HIQGLQSRYS ATQLHLHWGN PNDPHGSEHT VSGQHFAAEL HIVHYNSDLY PDASTASNKS EGLAVLAVLI EMGSFNPSYD KIFSHLQHVK YKGQEAFVPG FNIEELLPER TAEYYRYRGS LTTPPCNPTV LWTVFRNPVQ ISQEQLLALE TALYCTHMDD PSPREMINNF RQVQKFDERL VYTSFSQVQV CTAAGLS
Greater than 90% as determined by SDS-PAGE.
771
100 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers