missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Carbonic Anhydrase XII / CA12 Protein
A DNA sequence encoding the Homo sapiens (Human) Carbonic Anhydrase XII / CA12, was expressed in the hosts and tags indicated.
£183.90 - £1963.71
Specifications
Accession Number | NP_001209.1 |
---|---|
Gene ID (Entrez) | 771 |
Name | Carbonic Anhydrase XII / CA12 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15957439
|
enQuireBio™
QP5742-YE-100UG |
100 μg |
£467.00
100µg |
Estimated Shipment: 02-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15997439
|
enQuireBio™
QP5742-YE-500UG |
500 μg |
£1265.00
500µg |
Estimated Shipment: 02-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15967439
|
enQuireBio™
QP5742-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15987439
|
enQuireBio™
QP5742-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15907449
|
enQuireBio™
QP5742-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15977439
|
enQuireBio™
QP5742-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001209.1 | |
Carbonic Anhydrase XII / CA12 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
771 | |
Research Use Only | |
CA12 | |
Recombinant Protein | |
Yeast | |
APVNGSKWTY FGPDGENSWS KKYPSCGGLL QSPIDLHSDI LQYDASLTPL EFQGYNLSAN KQFLLTNNGH SVKLNLPSDM HIQGLQSRYS ATQLHLHWGN PNDPHGSEHT VSGQHFAAEL HIVHYNSDLY PDASTASNKS EGLAVLAVLI EMGSFNPSYD KIFSHLQHVK YKGQEAFVPG FNIEELLPER TAEYYRYRGS LTTPPCNPTV LWTVFRNPVQ ISQEQLLALE TALYCTHMDD PSPREMINNF RQVQKFDERL VYTSFSQVQV CTAAGLS | |
Greater than 90% as determined by SDS-PAGE. |