missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Caveolin-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Caveolin-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5766-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_001225.1 | |
Caveolin-3 Protein | |
Research Use Only | |
CAV3 | |
Recombinant Protein | |
E. coli | |
MMAEEHTDLE AQIVKDIHCK EIDLVNRDPK NINEDIVKVD FEDVIAEPVG TYSFDGVWKV SYTTFTVSKY WCYRLLSTLL GVPLALLWGF LFACISFCHI WAVVPCIKSY LIEIQCISHI YSLCIRTFCN PLFAALGQVC SSIKVVLRKE V | |
Greater than 80% as determined by SDS-PAGE. |
859 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |