missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD59 / CD59A / MAC-IP Protein
A DNA sequence encoding the Homo sapiens (Human) CD59 / CD59A / MAC-IP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5791-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_000602.1 | |
CD59 / CD59A / MAC-IP Protein | |
Research Use Only | |
CD59 | |
Recombinant Protein | |
E. coli | |
LQCYNCPNPT ADCKTAVNCS SDFDACLITK AGLQVYNKCW KFEHCNFNDV TTRLRENELT YYCCKKDLCN FNEQLEN | |
Greater than 90% as determined by SDS-PAGE. |
966 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |