missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD59 / CD59A / MAC-IP Protein
A DNA sequence encoding the Homo sapiens (Human) CD59 / CD59A / MAC-IP, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_000602.1 |
---|---|
Gene ID (Entrez) | 966 |
Name | CD59 / CD59A / MAC-IP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15957779
|
enQuireBio™
QP5791-ec-10ug |
10 μg | N/A | ||||||
15997779
|
enQuireBio™
QP5791-ec-50ug |
50 μg | N/A | ||||||
15947779
|
enQuireBio™
QP5791-ec-100ug |
100 μg |
£384.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15977779
|
enQuireBio™
QP5791-ec-200ug |
200 μg | N/A | ||||||
15987779
|
enQuireBio™
QP5791-ec-500ug |
500 μg | N/A | ||||||
15967779
|
enQuireBio™
QP5791-ec-1mg |
1 mg |
£1735.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_000602.1 | |
CD59 / CD59A / MAC-IP Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
966 | |
Research Use Only | |
CD59 | |
Recombinant Protein | |
E. coli | |
LQCYNCPNPT ADCKTAVNCS SDFDACLITK AGLQVYNKCW KFEHCNFNDV TTRLRENELT YYCCKKDLCN FNEQLEN | |
Greater than 90% as determined by SDS-PAGE. |