missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDNF Protein
A cDNA sequence encoding the CDNF was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10547-5ug
This item is not returnable.
View return policy
Specifications
CDNF Protein | |
Research Use Only | |
The ED50 as determined by its ability to stimulate the proliferation of rat C6 cells is 15-25microg/ml, corresponding to a specific activity of 40-67units/mg. | |
Human | |
Untagged | |
CDNF protein was lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4. |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
QEAGGRPGAD CEVCKEFLNR FYKSLIDRGV NFSLDTIEKE LISFCLDTKG KENRLCYYLG ATKDAATKIL SEVTRPMSVH MPAMKICEKL KKLDSQICEL KYEKTLDLAS VDLRKMRVAE LKQILHSWGE ECRACAEKTD YVNLIQELAP KYAATHPKTE L | |
Greater than 96.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |