missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDNF Protein
A cDNA sequence encoding the CDNF was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | CDNF Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The ED50 as determined by its ability to stimulate the proliferation of rat C6 cells is 15-25microg/ml, corresponding to a specific activity of 40-67units/mg. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974208
|
enQuireBio™
QP10547-5ug |
5 μg |
£99.15
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15964208
|
enQuireBio™
QP10547-20ug |
20 μg |
£155.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15954208
|
enQuireBio™
QP10547-1mg |
1 mg |
£2975.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CDNF Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
QEAGGRPGAD CEVCKEFLNR FYKSLIDRGV NFSLDTIEKE LISFCLDTKG KENRLCYYLG ATKDAATKIL SEVTRPMSVH MPAMKICEKL KKLDSQICEL KYEKTLDLAS VDLRKMRVAE LKQILHSWGE ECRACAEKTD YVNLIQELAP KYAATHPKTE L | |
Greater than 96.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
The ED50 as determined by its ability to stimulate the proliferation of rat C6 cells is 15-25microg/ml, corresponding to a specific activity of 40-67units/mg. | |
Human | |
Untagged | |
CDNF protein was lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4. |