missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CLEC4E Protein
A cDNA sequence encoding the CLEC4E was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11445-1mg
This item is not returnable.
View return policy
Specifications
CLEC4E Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MRCVVTFRIF QTCDEKKFQL PENFTELSCY NYGSGSVKNC CPLNWEYFQS SCYFFSTDTI SWALSLKNCS AMGAHLVVIN SQEEQEFLSY KKPKMREFFI GLSDQVVEGQ WQWVDGTPLT KSLSFWDVGE PNNIATLEDC ATMRDSSNPR QNWNDVTCFL NYFRICEMVG INPLNKGKSL | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CLEC4E protein solution (0.25 mg/ml) containing 20mM Tris 8.0 and 10% glycerol. |