missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CLIP-associating protein 1
A DNA sequence encoding the Homo sapiens(Human) CLIP-associating protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9972-iv-10ug
This item is not returnable.
View return policy
Specifications
NP_001193973.1 | |
CLIP-associating protein 1 | |
Research Use Only | |
CLASP2 | |
Recombinant Protein | |
E. coli | |
MRRLICKRIC DYKSFDDEES VDGNRPSSAA SAFKVPAPKT SGNPANSARK PGSAGGPKVG GASKEGGAGA VDEDDFIKAF TDVPSIQIYS SRELEETLNK IREILSDDKH DWDQRANALK KIRSLLVAGA AQYDCFFQHL RLLDGALKLS AKDLRSQVVR EACITVAHLS TVLGNKFDHG AEAIVPTLFN LVPNSAKVMA TSGCAAIRFI IRHTHVPRLI PLITSNCTSK SVPVRRRSFE FLDLLLQEWQ THSLERHAAV LVETIKKGIH DADAEARVEA RKTYMGLRNH FPGEAETLYN SLEPSYQKSL QTYLKSSGSV ASLPQSDRSS SSSQESLNRP FSSKWSTANP STVAGRVSAG SSKASSLPGS LQRSRSDIDV NAAAGAKAHH AAGQSVRSGR LGAGALNAGS YASLECEAFW RSGRTAKLYS V | |
Greater than 85% as determined by SDS-PAGE. |
23122 | |
10 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-B2M | |
Tris-based buffer, 50% glycerol |