missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CLIP-associating protein 1
A DNA sequence encoding the Homo sapiens(Human) CLIP-associating protein 2, was expressed in the hosts and tags indicated.
£428.07 - £1320.00
Specifications
Accession Number | NP_001193973.1 |
---|---|
Gene ID (Entrez) | 23122 |
Name | CLIP-associating protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16079111
|
enQuireBio™
QP9972-iv-10ug |
10 μg | N/A | ||||||
16089111
|
enQuireBio™
QP9972-iv-50ug |
50 μg | N/A | ||||||
16069111
|
enQuireBio™
QP9972-iv-100ug |
100 μg |
£1320.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001193973.1 | |
CLIP-associating protein 1 | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-B2M | |
Tris-based buffer, 50% glycerol |
23122 | |
Research Use Only | |
CLASP2 | |
Recombinant Protein | |
E. coli | |
MRRLICKRIC DYKSFDDEES VDGNRPSSAA SAFKVPAPKT SGNPANSARK PGSAGGPKVG GASKEGGAGA VDEDDFIKAF TDVPSIQIYS SRELEETLNK IREILSDDKH DWDQRANALK KIRSLLVAGA AQYDCFFQHL RLLDGALKLS AKDLRSQVVR EACITVAHLS TVLGNKFDHG AEAIVPTLFN LVPNSAKVMA TSGCAAIRFI IRHTHVPRLI PLITSNCTSK SVPVRRRSFE FLDLLLQEWQ THSLERHAAV LVETIKKGIH DADAEARVEA RKTYMGLRNH FPGEAETLYN SLEPSYQKSL QTYLKSSGSV ASLPQSDRSS SSSQESLNRP FSSKWSTANP STVAGRVSAG SSKASSLPGS LQRSRSDIDV NAAAGAKAHH AAGQSVRSGR LGAGALNAGS YASLECEAFW RSGRTAKLYS V | |
Greater than 85% as determined by SDS-PAGE. |