missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human CXCL8/IL-8 Protein
Recombinant Protein
Brand: Novus Biologicals™ NBP2-34905-100ug
463.20 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
Fully biologically active when compared to standard. The ED50 as determined by a chemotaxis bioassay using human CXCR2 transfected mouse BaF3 cells is less than 2 ng/mL, corresponding to a specific activity of >5.0 x 10^5 IU/mg.
Specifications
P10145, P10145, P10145 | |
PAGE, Bioactivity | |
3576 | |
100 μg | |
Less than 1 EU/ug of CXCL8/IL-8 as determined by LAL method. | |
CXCL8 | |
Angiogenesis, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
CXCL8/IL-8 |
LYOPH | |
Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. | |
8.4 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, IL-8/CXCL8, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C. | |
>97% pure by SDS-PAGE and HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Human CXCL8/IL-8 Protein > 100 μg
Spot an opportunity for improvement?Share a Content Correction