missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Defensin beta 2 Protein
Recombinant Protein
£103.00 - £2205.00
Specifications
Accession Number | O15263, O15263, O15263 |
---|---|
Concentration | LYOPH |
For Use With (Application) | PAGE, Bioactivity |
Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 130 mM NaCl. |
Gene ID (Entrez) | 13215 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18470922
|
Novus Biologicals™
NBP2-34910-5ug |
5 μg |
£103.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18432822
|
Novus Biologicals™
NBP2-34910-500ug |
500 μg |
£1270.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18434272
|
Novus Biologicals™
NBP2-34910-1mg |
1 mg |
£2205.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18450262
|
Novus Biologicals™
NBP2-34910-100ug |
100 μg |
£597.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Defensin beta 2 protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using immature human dendritic cells is in a concentration range of 10-100 ng/mL.Specifications
O15263, O15263, O15263 | |
PAGE, Bioactivity | |
13215 | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
BD-2, Defb2, Defb2 defensin beta 2, defensin beta 2 | |
Biologically Active Proteins | |
Defensin beta 2 |
LYOPH | |
Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 130 mM NaCl. | |
4.3 kDa | |
Less than 1 EU/ug of Defensin beta 2 as determined by LAL method. | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C. | |
>98% pure by SDS-PAGE and HPLC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title