missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DLST Protein
A cDNA sequence encoding the DLST was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11671-10ug
This item is not returnable.
View return policy
Specifications
DLST Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MLSRSRCVSR AFSRSLSAFQ KGNCPLGRRS LPGVSLCQGP GYPNSRKVVI NNSVFSVRFF RTTAVCKDDL VTVKTPAFAE SVTEGDVRWE KAVGDTVAED EVVCEIETDK TSVQVPSPAN GVIEALLVPD GGKVEGGTPL FTLRKTGAAP AKAKPAEAPA AAAPKAEPTA AAVPPPAAPI PTQMPPVPSP SQPPSGKPVS AVKPTVAPPL AEPGAGKGLR SEHREKMNRM RQRIAQRLKE AQNTCAMLTT FNEIDMSNIQ EMRARHKEAF LKKHNLKLGF MSAFVKASAF ALQEQPVVNA VIDDTTKEVV YRDYIDISVA VATPRGLVVP VIRNVEAMNF ADIERTITEL GEKARKNELA IEDMDGGTFT ISNGGVFGSL FGTPIINPPQ SAILGMHGIF DRPVAIGGKV EVRPMMYVAL TYDHRLIDGR EAVTFLRKIK AAVEDPRVLL LDL | |
DLST purity was found to be greater than 75% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
DLST (0.74 mg/ml) is supplied in 20mM HEPES buffer pH 8.0, 200mM NaCl and 20% glycerol. |