missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DLST Protein
A cDNA sequence encoding the DLST was constructed and used to recombinantly synthesize the protein.
£99.15 - £4515.00
Specifications
Name | DLST Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15987988
|
enQuireBio™
QP11671-2ug |
2 μg |
£99.15
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15967988
|
enQuireBio™
QP11671-10ug |
10 μg |
£155.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15977988
|
enQuireBio™
QP11671-1mg |
1 mg |
£4515.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
DLST Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
DLST (0.74 mg/ml) is supplied in 20mM HEPES buffer pH 8.0, 200mM NaCl and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MLSRSRCVSR AFSRSLSAFQ KGNCPLGRRS LPGVSLCQGP GYPNSRKVVI NNSVFSVRFF RTTAVCKDDL VTVKTPAFAE SVTEGDVRWE KAVGDTVAED EVVCEIETDK TSVQVPSPAN GVIEALLVPD GGKVEGGTPL FTLRKTGAAP AKAKPAEAPA AAAPKAEPTA AAVPPPAAPI PTQMPPVPSP SQPPSGKPVS AVKPTVAPPL AEPGAGKGLR SEHREKMNRM RQRIAQRLKE AQNTCAMLTT FNEIDMSNIQ EMRARHKEAF LKKHNLKLGF MSAFVKASAF ALQEQPVVNA VIDDTTKEVV YRDYIDISVA VATPRGLVVP VIRNVEAMNF ADIERTITEL GEKARKNELA IEDMDGGTFT ISNGGVFGSL FGTPIINPPQ SAILGMHGIF DRPVAIGGKV EVRPMMYVAL TYDHRLIDGR EAVTFLRKIK AAVEDPRVLL LDL | |
DLST purity was found to be greater than 75% as determined by SDS-PAGE. |