missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GADD45B Protein
A cDNA sequence encoding the GADD45B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11920-10ug
This item is not returnable.
View return policy
Specifications
GADD45B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MTLEELVACD NAAQKMQTVT AAVEELLVAA QRQDRLTVGV YESAKLMNVD PDSVVLCLLA IDEEEEDDIA LQIHFTLIQS FCCDNDINIVRVSGMQRLAQ LLGEPAETQG TTEARDLHCL LVTNPHTDAW KSHGLVEVAS YCEESRGNNQ WVPYISLQER | |
Greater than 95.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The GADD45B protein solution contains 20mM Tris-HCl pH 7.5 and 20% glycerol. |