missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GADD45B Protein
A cDNA sequence encoding the GADD45B was constructed and used to recombinantly synthesize the protein.
£99.15 - £1985.00
Specifications
Name | GADD45B Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15968798
|
enQuireBio™
QP11920-10ug |
10 μg |
£99.15
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15988798
|
enQuireBio™
QP11920-50ug |
50 μg |
£155.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15978798
|
enQuireBio™
QP11920-1mg |
1 mg |
£1985.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
GADD45B Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The GADD45B protein solution contains 20mM Tris-HCl pH 7.5 and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MTLEELVACD NAAQKMQTVT AAVEELLVAA QRQDRLTVGV YESAKLMNVD PDSVVLCLLA IDEEEEDDIA LQIHFTLIQS FCCDNDINIVRVSGMQRLAQ LLGEPAETQG TTEARDLHCL LVTNPHTDAW KSHGLVEVAS YCEESRGNNQ WVPYISLQER | |
Greater than 95.0% as determined by SDS-PAGE. |