missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF-15 Protein
A DNA sequence encoding the Homo sapiens (Human) GDF-15, was expressed in the hosts and tags indicated.
£246.00 - £1780.00
Specifications
Accession Number | NP_004855.2 |
---|---|
Gene ID (Entrez) | 9518 |
Name | GDF-15 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15975629
|
enQuireBio™
QP4998-ye-50ug |
50 μg |
£246.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15935629
|
enQuireBio™
QP4998-ye-100ug |
100 μg |
£393.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15955629
|
enQuireBio™
QP4998-ye-200ug |
200 μg |
£635.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15965629
|
enQuireBio™
QP4998-ye-500ug |
500 μg |
£1095.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15945629
|
enQuireBio™
QP4998-ye-1mg |
1 mg |
£1780.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_004855.2 | |
GDF-15 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
9518 | |
Research Use Only | |
GDF15 | |
Recombinant Protein | |
Yeast | |
RNGDHCPLGP GRCCRLHTVR ASLEDLGWAD WVLSPREVQV TMCIGACPSQ FRAANMHAQI KTSLHRLKPD TVPAPCCVPA SYNPMVLIQK TDTGVSLQTY DDLLAKDCHC I | |
Greater than 90% as determined by SDS-PAGE. |