missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GET4 Protein
A cDNA sequence encoding the GET4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11953-1mg
This item is not returnable.
View return policy
Specifications
GET4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAAAAM AEQESARNGG RNRGGVQRVE GKLRASVEKG DYYEAHQMYR TLFFRYMSQS KHTEARELMY SGALLFFSHG QQNSAADLSM LVLESLEKAE VEVADELLEN LAKVFSLMDP NSPERVTFVS RALKWSSGGS GKLGHPRLHQ LLALTLWKEQ NYCESRYHFL HSADGEGCAN MLVEYSTSRG FRSEVDMFVA QAVLQFLCLK NKSSASVVFT TYTQKHPSIE DGPPFVEPLL NFIWFLLLAV DGGKLTVFTV LCEQYQPSLR RDPMYNEYLD RIGQLFFGVP PKQTSSYGGL LGNLLTSLMG SSEQEDGEES PSDGSPIELD | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GET4 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |